Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyruvate Assay Kit suffici
Catalog No. 502904604
Encompass
Pyruvate is a key intermediate in cellular metabolic pathways. Pyruvate can be converted to carbohydrates via gluconeogenesis to fatty acids or energy through acetyl-CoA to the amino acid alanine and to ethanol. Abnormal levels of pyruvate have been linked to liver diseases and metabolic disorders. Simple direct and automation-ready procedures for measuring pyruvate concentrations find wide applications in research and drug discovery.
Catalog No. 50-290-4604 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3321KT
Pyruvate is a key intermediate in cellular metabolic pathways. Pyruvate can be converted to carbohydrates via gluconeogenesis to fatty acids or energy through acetyl-CoA to the amino acid alanine and to ethanol. Abnormal levels of pyruvate have been linked to liver diseases and metabolic disorders. Simple direct and automation-ready procedures for measuring pyruvate concentrations find wide applications in research and drug discovery.