Learn More
Medchemexpress LLC Pyruvate carboxylase-IN-1 | 70205-50-4 | 99.4% | 242.27 g/mol | C15H14O3 | 5 MG
SDP
Catalog No. 5000221464
Encompass_Preferred
Pyruvate Carboxylase-IN-1 is a small-molecule inhibitor of pyruvate carboxylase with reported antiproliferative activity. It shows submicromolar cellular potency and is supplied as a high-purity research reagent for biochemical and cell-based studies.
  • High purity (99.4%) for reliable experimental reproducibility.
  • Submicromolar potency in cellular assays (IC50 ≈ 0.104 μM) and low-micromolar activity in lysates (IC50 ≈ 0.204 μM).
  • Suitable for biochemical and cell-based assays targeting mitochondrial metabolism.
  • Molecular weight 242.27 g/mol and formula C15H14O3 for accurate mass calculation.
  • Available in small research pack sizes suitable for screening and validation.
Catalog No. 50-002-21464 Supplier Medchemexpress LLC Supplier No. HY1426845MG
Pyruvate Carboxylase-IN-1 is a small-molecule inhibitor of pyruvate carboxylase with reported antiproliferative activity. It shows submicromolar cellular potency and is supplied as a high-purity research reagent for biochemical and cell-based studies.
  • High purity (99.4%) for reliable experimental reproducibility.
  • Submicromolar potency in cellular assays (IC50 ≈ 0.104 μM) and low-micromolar activity in lysates (IC50 ≈ 0.204 μM).
  • Suitable for biochemical and cell-based assays targeting mitochondrial metabolism.
  • Molecular weight 242.27 g/mol and formula C15H14O3 for accurate mass calculation.
  • Available in small research pack sizes suitable for screening and validation.