Learn More
Medchemexpress LLC Pyruvate Carboxylase-IN-1 | 70205-50-4 | 99.4% | C15H14O3 | 50 MG
SDP
Catalog No. 5000353684
Encompass_Preferred
Pyruvate Carboxylase-IN-1 (compound 37) is a potent pyruvate carboxylase inhibitor with antiproliferative activity. The IC50 values ​​of pyruvate carboxylase-IN-1 against PC in cell lysates and cells are 0.204 and 0.104 μM, respectively.
  • Potent pyruvate carboxylase inhibitor.
  • Exhibits antiproliferative activity.
  • IC50 value against PC in cell lysates: 0.204 μM.
  • IC50 value against PC in cells: 0.104 μM.
Catalog No. 50-003-53684 Supplier Medchemexpress LLC Supplier No. HY14268450MG
Pyruvate Carboxylase-IN-1 (compound 37) is a potent pyruvate carboxylase inhibitor with antiproliferative activity. The IC50 values ​​of pyruvate carboxylase-IN-1 against PC in cell lysates and cells are 0.204 and 0.104 μM, respectively.
  • Potent pyruvate carboxylase inhibitor.
  • Exhibits antiproliferative activity.
  • IC50 value against PC in cell lysates: 0.204 μM.
  • IC50 value against PC in cells: 0.104 μM.