Learn More
Medchemexpress LLC Pyruvate Carboxylase-IN-5 | 3047101-14-1 | 98.9% | 294.24 | 25 MG
SDP
Catalog No. 5000393260
Encompass_Preferred
Pyruvate Carboxylase-IN-5 (compound 6m) is a highly selective and permeable inhibitor of pyruvate carboxylase. This mitochondrial enzyme plays a crucial role in catalyzing the carboxylation of pyruvate to oxaloacetate, a process essential for maintaining steady-state levels of Krebs cycle intermediates. These intermediates are vital precursors for the synthesis of biomacromolecules such as amino acids, fatty acids, and glucose.
  • High selectivity and permeability
  • Inhibits pyruvate carboxylase
  • Supports research into metabolism and biosynthesis pathways
Catalog No. 50-003-93260 Supplier Medchemexpress LLC Supplier No. 25MGHY163771
Pyruvate Carboxylase-IN-5 (compound 6m) is a highly selective and permeable inhibitor of pyruvate carboxylase. This mitochondrial enzyme plays a crucial role in catalyzing the carboxylation of pyruvate to oxaloacetate, a process essential for maintaining steady-state levels of Krebs cycle intermediates. These intermediates are vital precursors for the synthesis of biomacromolecules such as amino acids, fatty acids, and glucose.
  • High selectivity and permeability
  • Inhibits pyruvate carboxylase
  • Supports research into metabolism and biosynthesis pathways