Learn More
Medchemexpress LLC Pyruvate Carboxylase-IN-5 | 3047101-14-1 | 98.9% | 294.24 | 5 MG
SDP
Catalog No. 5000391011
Encompass_Preferred
Pyruvate Carboxylase-IN-5 (compound 6m) is a highly selective and permeable inhibitor of pyruvate carboxylase. Pyruvate carboxylase is a mitochondrial enzyme that catalyzes the carboxylation of pyruvate to oxaloacetate. This process is crucial for maintaining steady-state levels of Krebs cycle intermediates, which are precursors for the synthesis of biomacromolecules like amino acids, fatty acids, and glucose.
  • High selectivity and permeability.
  • Inhibitor of pyruvate carboxylase.
  • Plays an important role in maintaining steady-state levels of Krebs cycle intermediates.
  • Research use only.
Catalog No. 50-003-91011 Supplier Medchemexpress LLC Supplier No. HY1637715MG
Pyruvate Carboxylase-IN-5 (compound 6m) is a highly selective and permeable inhibitor of pyruvate carboxylase. Pyruvate carboxylase is a mitochondrial enzyme that catalyzes the carboxylation of pyruvate to oxaloacetate. This process is crucial for maintaining steady-state levels of Krebs cycle intermediates, which are precursors for the synthesis of biomacromolecules like amino acids, fatty acids, and glucose.
  • High selectivity and permeability.
  • Inhibitor of pyruvate carboxylase.
  • Plays an important role in maintaining steady-state levels of Krebs cycle intermediates.
  • Research use only.