Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyruvate decarboxylase | 9001-04-1 | ≥1 U/mg solid | 60 kDa | 20 U
SDP

Catalog No. 5000398710
Encompass_Preferred

Pyruvate decarboxylase (PDC) is an enzyme that catalyzes the decarboxylation of pyruvic acid to acetaldehyde. It specifically catalyzes the non-oxidative conversion of pyruvate (or other 2-oxo acids) to acetaldehyde and CO2. This enzyme is a key component in alcoholic fermentation and is found in yeast, some bacteria, and various plants.

  • Appearance: Solid
  • Color: White to off-white
  • EC Number: 4.1.1.1
  • Unit definition: One unit converts 1 μmol of pyruvic acid to acetaldehyde per minute at pH 6.0, 25°C.

Catalog No. 50-003-98710 Supplier Medchemexpress LLC Supplier No. HYP279620U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyruvate decarboxylase (PDC) is an enzyme that catalyzes the decarboxylation of pyruvic acid to acetaldehyde. It specifically catalyzes the non-oxidative conversion of pyruvate (or other 2-oxo acids) to acetaldehyde and CO2. This enzyme is a key component in alcoholic fermentation and is found in yeast, some bacteria, and various plants.

  • Appearance: Solid
  • Color: White to off-white
  • EC Number: 4.1.1.1
  • Unit definition: One unit converts 1 μmol of pyruvic acid to acetaldehyde per minute at pH 6.0, 25°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.