Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyruvate Dehydrogenase E1 beta subunit Antibody (YA114) | Predicted band size: 39 kDa; Observed band size: 34 kDa | 10UL
SDP

Catalog No. 5000395736
Encompass_Preferred

Pyruvate Dehydrogenase E1 beta subunit Antibody (YA114) is a rabbit-derived monoclonal antibody targeting the Pyruvate Dehydrogenase E1 beta subunit. This subunit is a key component of the pyruvate dehydrogenase complex, which catalyzes the overall conversion of pyruvate to acetyl-CoA and CO2, thereby linking the glycolytic pathway to the tricarboxylic acid cycle. This product is intended for use as laboratory chemicals and in the manufacture of substances.

  • Host is rabbit
  • Reacts with human, mouse, and rat samples
  • Non-conjugated
  • Predicted band size: 39 kDa
  • Observed band size: 34 kDa
  • Protein A affinity purified
  • Formulated with 0.05% BSA and 40% glycerol
  • Store at -20°C for 1 year
  • Liquid appearance
  • Suitable for Western Blot (WB)
  • Suitable for Immunohistochemistry (IHC-P)
  • Suitable for Flow Cytometry (FC)
  • Suitable for Immunoprecipitation (IP)

Catalog No. 50-003-95736 Supplier Medchemexpress LLC Supplier No. HYP8029110UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyruvate Dehydrogenase E1 beta subunit Antibody (YA114) is a rabbit-derived monoclonal antibody targeting the Pyruvate Dehydrogenase E1 beta subunit. This subunit is a key component of the pyruvate dehydrogenase complex, which catalyzes the overall conversion of pyruvate to acetyl-CoA and CO2, thereby linking the glycolytic pathway to the tricarboxylic acid cycle. This product is intended for use as laboratory chemicals and in the manufacture of substances.

  • Host is rabbit
  • Reacts with human, mouse, and rat samples
  • Non-conjugated
  • Predicted band size: 39 kDa
  • Observed band size: 34 kDa
  • Protein A affinity purified
  • Formulated with 0.05% BSA and 40% glycerol
  • Store at -20°C for 1 year
  • Liquid appearance
  • Suitable for Western Blot (WB)
  • Suitable for Immunohistochemistry (IHC-P)
  • Suitable for Flow Cytometry (FC)
  • Suitable for Immunoprecipitation (IP)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.