Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyruvate Dehydrogenase E1 beta subunit Antibody (YA114) | Protein A affinity purified | Predicted: 39 kDa; Observed: 34 kDa | 50 UL
SDP

Catalog No. 5000397881
Encompass_Preferred

Pyruvate Dehydrogenase E1 beta subunit Antibody (YA114) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting the Pyruvate Dehydrogenase E1 beta subunit. This subunit is a key component of the pyruvate dehydrogenase complex, which catalyzes the conversion of pyruvate to acetyl-CoA and CO₂, linking the glycolytic pathway to the tricarboxylic acid cycle.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Research field: Signal transduction
  • Purity: Protein A affinity purified
  • Molecular weight: Predicted 39 kDa, observed 34 kDa
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Applications: WB (1:500-1:2000), IHC-P (1:50-1:200), FC (1:50-1:100), IP (assay dependent)

Catalog No. 50-003-97881 Supplier Medchemexpress LLC Supplier No. HYP8029150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyruvate Dehydrogenase E1 beta subunit Antibody (YA114) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting the Pyruvate Dehydrogenase E1 beta subunit. This subunit is a key component of the pyruvate dehydrogenase complex, which catalyzes the conversion of pyruvate to acetyl-CoA and CO₂, linking the glycolytic pathway to the tricarboxylic acid cycle.

  • Host: Rabbit
  • Reactivity: Human, mouse, rat
  • Conjugation: Non-conjugated
  • Research field: Signal transduction
  • Purity: Protein A affinity purified
  • Molecular weight: Predicted 39 kDa, observed 34 kDa
  • Formulation: Supplied in 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol with 0.05% Sodium Azide
  • Storage: Stored at -20°C for 1 year, avoid repeated freeze/thaw cycles
  • Applications: WB (1:500-1:2000), IHC-P (1:50-1:200), FC (1:50-1:100), IP (assay dependent)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.