Learn More
Medchemexpress LLC Pyruvate dehydrogenase E2 antibody (YA912) | 10 UL
SDP
Catalog No. 5000442589
Encompass_Preferred
Pyruvate Dehydrogenase E2 Antibody (YA912) is a mouse-derived, non-conjugated IgG1 monoclonal antibody. It targets Pyruvate Dehydrogenase E2 and is intended for research use only. The antibody is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, with a pH of 7.3.
  • Derived from mouse
  • Monoclonal antibody
  • Non-conjugated IgG1 isotype
  • Targets pyruvate dehydrogenase E2
  • Suitable for Western Blot, Immunocytochemistry/Immunofluorescence, and Immunoprecipitation applications
  • Host reactivity includes human and mouse
  • Stored at -20°C for 1 year
  • Predicted and observed band size of 69 kDa
Catalog No. 50-004-42589 Supplier Medchemexpress LLC Supplier No. HYP8107010UL
Pyruvate Dehydrogenase E2 Antibody (YA912) is a mouse-derived, non-conjugated IgG1 monoclonal antibody. It targets Pyruvate Dehydrogenase E2 and is intended for research use only. The antibody is supplied in PBS containing 50% glycerol, 0.5% BSA, and 0.02% sodium azide, with a pH of 7.3.
  • Derived from mouse
  • Monoclonal antibody
  • Non-conjugated IgG1 isotype
  • Targets pyruvate dehydrogenase E2
  • Suitable for Western Blot, Immunocytochemistry/Immunofluorescence, and Immunoprecipitation applications
  • Host reactivity includes human and mouse
  • Stored at -20°C for 1 year
  • Predicted and observed band size of 69 kDa