Learn More
Apexbio Technology LLC Pyruvate-DH-PDH-Act-Assay-Kit
SDP
Catalog No. 5000567735
Encompass_Preferred
Pyruvate Dehydrogenase (PDH) Activity Assay Kit is a pivotal tool in biomedical research designed to measure the activity of the PDH enzyme complex integral to the Krebs cycle Originating from the need to study metabolic pathways it evaluates the conversion of pyruvate to acetyl-CoA a crucial step in cellular respiration This assay is widely used in in vitro studies with activity commonly detected in the nanomolar to low micromolar range It serves typical applications such as assessing mitochondrial function in cell models evaluating metabolic responses in animal studies and high-throughput drug screening Experimental concentrations typically depend on the specific research design ensuring versatility across various experimental settings This comprehensive kit is essential for researchers focusing on cellular energy metabolism and its related disorders
Catalog No. 50-005-67735 Supplier Apexbio Technology LLC Supplier No. K221896
Pyruvate Dehydrogenase (PDH) Activity Assay Kit is a pivotal tool in biomedical research designed to measure the activity of the PDH enzyme complex integral to the Krebs cycle Originating from the need to study metabolic pathways it evaluates the conversion of pyruvate to acetyl-CoA a crucial step in cellular respiration This assay is widely used in in vitro studies with activity commonly detected in the nanomolar to low micromolar range It serves typical applications such as assessing mitochondrial function in cell models evaluating metabolic responses in animal studies and high-throughput drug screening Experimental concentrations typically depend on the specific research design ensuring versatility across various experimental settings This comprehensive kit is essential for researchers focusing on cellular energy metabolism and its related disorders