Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyruvate Kinase from rabbi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P913625KU
Encompass
Pyruvate Kinase (PK) is a glycolysis enzyme and has four isozymes L R M1 and M2. The L isozyme is localized in gluconeogenic tissues particularly in the liver. Whereas the R and M1 are localized in the adult skeletal muscles heart brain and erythrocytes respectively. M2 is localized in the nucleus of the cells.
Catalog No. 50-290-2636