Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyruvate Kinase from rabbi
Catalog No. 502902484
Encompass
Pyruvate kinase from rabbit muscle catalyzes an ATP-dependent phosphorylation of glycolate to yield 2-phosphoglycolate.
Catalog No. 50-290-2484 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P150625KU
Pyruvate kinase from rabbit muscle catalyzes an ATP-dependent phosphorylation of glycolate to yield 2-phosphoglycolate.