Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyruvate Kinase from rabbi
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P15065KU
Encompass
Pyruvate kinase from rabbit muscle catalyzes an ATP-dependent phosphorylation of glycolate to yield 2-phosphoglycolate.
Catalog No. 50-289-9495