Learn More
Medchemexpress LLC Pyruvate Kinase, Microorganism | 9001-59-6 | 237 kDa | 1 KU
SDP
Catalog No. 5000398619
Encompass_Preferred
Pyruvate Kinase, Microorganism (PK) is a glycolytic enzyme that catalyzes the conversion of phosphoenolpyruvate and ADP to pyruvate and ATP. This product is for research use only.
  • Catalyzes conversion of phosphoenolpyruvate and ADP to pyruvate and ATP
  • Theoretical molecular weight: 237 kDa
  • Optimum pH: 7.0-7.5
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-003-98619 Supplier Medchemexpress LLC Supplier No. HYP28061KU
Pyruvate Kinase, Microorganism (PK) is a glycolytic enzyme that catalyzes the conversion of phosphoenolpyruvate and ADP to pyruvate and ATP. This product is for research use only.
  • Catalyzes conversion of phosphoenolpyruvate and ADP to pyruvate and ATP
  • Theoretical molecular weight: 237 kDa
  • Optimum pH: 7.0-7.5
  • Appears as a white to off-white solid
  • For research use only