Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyruvate Kinase PK from ra
Catalog No. 502900370
Encompass
ATPpyruvate 2-O-phosphotransferasePyruvate kinase has a molar mass of 237 000 and exists as a tetramer. Each polypeptide chain of this tetramer has a molar mass of 57 200. The enzyme contains two identical catalytic particles called protomers. Each of these protomers contains two polypeptide chains. Each protomer contains one site each for Mn2 and phosphoenolpyruvate.
Catalog No. 50-290-0370 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10128155001
ATPpyruvate 2-O-phosphotransferasePyruvate kinase has a molar mass of 237 000 and exists as a tetramer. Each polypeptide chain of this tetramer has a molar mass of 57 200. The enzyme contains two identical catalytic particles called protomers. Each of these protomers contains two polypeptide chains. Each protomer contains one site each for Mn2 and phosphoenolpyruvate.