Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyruvate Kinase, rabbit muscle, Thermo Scientific Chemicals

Catalog No. AAJ61373EXV
Change view
Click to view available options
Quantity:
50 kU
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ61373EXV 50 kU Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61373EXV Supplier Thermo Scientific Chemicals Supplier No. J61373EXV

 

Pyruvate kinase has been used in the preparation of cell free extract from BL21 CP strain of E. coli culture for highly productive cell-free protein expression. The enzyme has been used to assay the pyruvate kinase activity by incubating with eluted DAPk (Death-associated protein kinase), a serine/threonine kinase that binds and activates pyruvate kinase. It is involved in glycolysis and in gluconeogenesis. It has been used in PGM enzyme assays to determine phosphoglycerate mutase activity. It is also used to study pyruvate kinase (PK) deficiency.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9001-59-6
MDL Number MFCD00081946
Synonym Phosphoenolpyruvate kinase,Phosphoenol transphosphorylase

Specifications

Packaging Plastic bottle
Quantity 50 kU
Solubility Information Soluble in water ( 1mg/ml).
Physical Form Lyophilized Powder
Chemical Name or Material Pyruvate Kinase
EINECSNumber 232-616-2
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.