Learn More
Medchemexpress LLC Pyruvic-1-13C acid | 99124-30-8 | MFCD10566986 | 98.7% | 89.05 | C213CH4O3 | 5mg
SDP
Catalog No. 5000268593
Encompass_Preferred
Pyruvic acid-13C is the 13C labeled Pyruvic acid[1] Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats[2]
Catalog No. 50-002-68593 Supplier Medchemexpress LLC Supplier No. HYY0781S15MG
Pyruvic acid-13C is the 13C labeled Pyruvic acid[1] Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats[2]