Learn More
Medchemexpress LLC Pyruvic acid | 127-17-3 | MFCD00002585 | 88.06 | C3H4O3 | 25mg
SDP
Catalog No. 5000268591
Encompass_Preferred
Pyruvic acid (Standard) is the analytical standard of Pyruvic acid This product is intended for research and analytical applications Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats
Catalog No. 50-002-68591 Supplier Medchemexpress LLC Supplier No. HYY0781R25MG
Pyruvic acid (Standard) is the analytical standard of Pyruvic acid This product is intended for research and analytical applications Pyruvic acid is an intermediate metabolite in the metabolism of carbohydrates proteins and fats