Learn More
Selleck Chemical LLC Pyruvic acid S3143-100mg
SDP
Catalog No. 503109379
Encompass_Preferred
Pyruvic acid (Acetylformic acid) is an important organic chemical intermediate that plays a role in cardiomyocyte pathophysiology and therapy Pyruvic acid markedly increases the level of lactate dehydrogenase (LDH) and creatine kinase (CK) and reduces the level of Ca2 Mg2 -ATPase and Na K -ATPase
Catalog No. 50-310-9379 Supplier Selleck Chemical LLC Supplier No. S3143100MG
Pyruvic acid (Acetylformic acid) is an important organic chemical intermediate that plays a role in cardiomyocyte pathophysiology and therapy Pyruvic acid markedly increases the level of lactate dehydrogenase (LDH) and creatine kinase (CK) and reduces the level of Ca2 Mg2 -ATPase and Na K -ATPase