Learn More
Alkali Scientific Pyruvic Acid Sodium Salt, [Sodium Pyruvate], 1 Kg
SDP
Catalog No. 5000661334
Encompass_Preferred
Sodium Pyruvate (Pyruvic Acid Sodium Salt) is a key reagent used in metabolic studies and cell culture applications. This 1kg pack is ideal for laboratories requiring a large volume of sodium pyruvate for extensive cell culture or biochemical research. Sodium pyruvate is a key intermediate in the glycolytic pathway and is used to maintain optimal metabolic conditions for mammalian cells. It supports cellular respiration and enhances cell growth, particularly in high-density cultures. This compound is commonly added to culture media to improve cell viability, protect against oxidative damage, and support cellular energy production. It is also used in studies of mitochondrial function and metabolism.
Catalog No. 50-006-61334 Supplier Alkali Scientific Supplier No. PYR002CP
Sodium Pyruvate (Pyruvic Acid Sodium Salt) is a key reagent used in metabolic studies and cell culture applications. This 1kg pack is ideal for laboratories requiring a large volume of sodium pyruvate for extensive cell culture or biochemical research. Sodium pyruvate is a key intermediate in the glycolytic pathway and is used to maintain optimal metabolic conditions for mammalian cells. It supports cellular respiration and enhances cell growth, particularly in high-density cultures. This compound is commonly added to culture media to improve cell viability, protect against oxidative damage, and support cellular energy production. It is also used in studies of mitochondrial function and metabolism.