Learn More
Alkali Scientific Pyruvic Acid Sodium Salt, [Sodium Pyruvate], 2.5 Kg
SDP
Catalog No. 5000661203
Encompass_Preferred
Sodium pyruvate (pyruvic acid sodium salt) is a critical metabolic intermediate and buffer component used in cell culture applications to support cellular growth and function. This 2.5 kg quantity is ideal for large-scale research and cell culture production, ensuring an adequate supply for routine and specialized applications. Sodium pyruvate is vital for maintaining cellular energy balance by facilitating glycolysis and other metabolic pathways. It stabilizes pH in culture media, promotes cell survival, and enhances cellular proliferation. This form of pyruvate is widely used in experimental studies focusing on metabolism, enzymatic reactions, and as a supplement in cell culture mediums for improved cell viability.
Catalog No. 50-006-61203 Supplier Alkali Scientific Supplier No. PYR005CP
Sodium pyruvate (pyruvic acid sodium salt) is a critical metabolic intermediate and buffer component used in cell culture applications to support cellular growth and function. This 2.5 kg quantity is ideal for large-scale research and cell culture production, ensuring an adequate supply for routine and specialized applications. Sodium pyruvate is vital for maintaining cellular energy balance by facilitating glycolysis and other metabolic pathways. It stabilizes pH in culture media, promotes cell survival, and enhances cellular proliferation. This form of pyruvate is widely used in experimental studies focusing on metabolism, enzymatic reactions, and as a supplement in cell culture mediums for improved cell viability.