Learn More
Alkali Scientific Pyruvic Acid Sodium Salt, [Sodium Pyruvate], 25 G
SDP
Catalog No. 5000661608
Encompass_Preferred
Sodium pyruvate, also known as pyruvic acid sodium salt, is a crucial metabolic intermediate commonly used in cell culture applications. This 25g quantity is particularly useful in research where precise quantities are required for experimentation and medium preparation. Sodium pyruvate serves as a buffer component that helps maintain the pH of the culture medium while providing a carbon source for cellular metabolism. It is essential in supporting the growth and function of mammalian cells, including primary cultures and cell lines. Additionally, sodium pyruvate can serve as an energy substrate in metabolic studies and tissue culture environments to promote cellular viability.
Catalog No. 50-006-61608 Supplier Alkali Scientific Supplier No. PYR004CP
Sodium pyruvate, also known as pyruvic acid sodium salt, is a crucial metabolic intermediate commonly used in cell culture applications. This 25g quantity is particularly useful in research where precise quantities are required for experimentation and medium preparation. Sodium pyruvate serves as a buffer component that helps maintain the pH of the culture medium while providing a carbon source for cellular metabolism. It is essential in supporting the growth and function of mammalian cells, including primary cultures and cell lines. Additionally, sodium pyruvate can serve as an energy substrate in metabolic studies and tissue culture environments to promote cellular viability.