Learn More
Alkali Scientific Pyruvic Acid Sodium Salt, [Sodium Pyruvate], 5 Kg
SDP
Catalog No. 5000661210
Encompass_Preferred
Sodium pyruvate, or pyruvic acid sodium salt, is a critical component used in cell culture as a buffer and metabolic intermediate. This 5 kg quantity is used in large-scale research facilities or production labs where continuous or high-throughput cell culture applications are required. Sodium pyruvate helps maintain the pH of cell culture media and provides an energy source for cells, particularly in mammalian cell cultures. It plays a significant role in cellular metabolism by fueling glycolysis and supporting mitochondrial function. Sodium pyruvate is widely used in experimental studies, metabolic assays, and for the maintenance of cell lines under various growth conditions, including long-term culture.
Catalog No. 50-006-61210 Supplier Alkali Scientific Supplier No. PYR008CP
Sodium pyruvate, or pyruvic acid sodium salt, is a critical component used in cell culture as a buffer and metabolic intermediate. This 5 kg quantity is used in large-scale research facilities or production labs where continuous or high-throughput cell culture applications are required. Sodium pyruvate helps maintain the pH of cell culture media and provides an energy source for cells, particularly in mammalian cell cultures. It plays a significant role in cellular metabolism by fueling glycolysis and supporting mitochondrial function. Sodium pyruvate is widely used in experimental studies, metabolic assays, and for the maintenance of cell lines under various growth conditions, including long-term culture.