Learn More
Alkali Scientific Pyruvic Acid Sodium Salt, [Sodium Pyruvate], 50 G
SDP
Catalog No. 5000661396
Encompass_Preferred
Sodium pyruvate, also known as pyruvic acid sodium salt, is a key buffer component and metabolic intermediate commonly used in cell culture applications. This 50g quantity is typically used for laboratory experiments and medium preparation where a smaller quantity is required for controlled studies. Sodium pyruvate helps maintain pH levels in cell culture media, providing stability and energy support for cells. It plays a crucial role in cellular metabolism by participating in glycolysis and various biochemical processes. This reagent is commonly utilized in laboratory studies involving mammalian cells, stem cells, and metabolic research to promote cell growth, viability, and function under experimental conditions.
Catalog No. 50-006-61396 Supplier Alkali Scientific Supplier No. PYR006CP
Sodium pyruvate, also known as pyruvic acid sodium salt, is a key buffer component and metabolic intermediate commonly used in cell culture applications. This 50g quantity is typically used for laboratory experiments and medium preparation where a smaller quantity is required for controlled studies. Sodium pyruvate helps maintain pH levels in cell culture media, providing stability and energy support for cells. It plays a crucial role in cellular metabolism by participating in glycolysis and various biochemical processes. This reagent is commonly utilized in laboratory studies involving mammalian cells, stem cells, and metabolic research to promote cell growth, viability, and function under experimental conditions.