Learn More
Alkali Scientific Pyruvic Acid Sodium Salt, [Sodium Pyruvate], 500 G
SDP
Catalog No. 5000661224
Encompass_Preferred
Sodium pyruvate (pyruvic acid sodium salt) is a widely used reagent in biological research, particularly in cell culture applications. This 500g quantity is ideal for laboratories with high-volume research or production needs, where larger amounts of pyruvate are necessary. Sodium pyruvate acts as a buffer component that stabilizes pH in culture media and also provides an essential energy substrate for cultured cells. It supports cellular metabolism, promotes cell growth, and is used extensively in experiments studying mitochondrial function and glycolysis. Sodium pyruvate is essential for maintaining optimal conditions in cell culture environments, ensuring cell viability and proper cellular function during extended incubation periods.
Catalog No. 50-006-61224 Supplier Alkali Scientific Supplier No. PYR007CP
Sodium pyruvate (pyruvic acid sodium salt) is a widely used reagent in biological research, particularly in cell culture applications. This 500g quantity is ideal for laboratories with high-volume research or production needs, where larger amounts of pyruvate are necessary. Sodium pyruvate acts as a buffer component that stabilizes pH in culture media and also provides an essential energy substrate for cultured cells. It supports cellular metabolism, promotes cell growth, and is used extensively in experiments studying mitochondrial function and glycolysis. Sodium pyruvate is essential for maintaining optimal conditions in cell culture environments, ensuring cell viability and proper cellular function during extended incubation periods.