Learn More
Medchemexpress LLC Pyruvic aldehyde | 78-98-8 | MFCD00006960 | 97.3% | 72.06 g·mol⁻¹ | C3H4O2 | 1 ML
SDP
Catalog No. 5000300914
Encompass_Preferred
Pyruvic aldehyde is supplied as a 40% (w/w) aqueous solution of the reactive dicarbonyl 2-oxopropanal. It is used as a reagent in organic synthesis and chemical research, and appears in studies of flavoring agents and tanning processes. The solution form reduces handling of the neat aldehyde; users should follow the supplier safety data sheet for storage, transport, and handling requirements.
  • 40% aqueous solution for simplified handling and reduced vapor hazards.
  • Reactive dicarbonyl reagent for organic synthesis and analytical studies.
  • Applicable to flavoring agent research and tanning process investigations.
  • Molecular weight 72.06 g·mol⁻¹ and formula C3H4O2 aid analytical identification.
  • Refer to the safety data sheet for storage, handling, and hazard information.
Catalog No. 50-003-00914 Supplier Medchemexpress LLC Supplier No. HYW0200141ML
Pyruvic aldehyde is supplied as a 40% (w/w) aqueous solution of the reactive dicarbonyl 2-oxopropanal. It is used as a reagent in organic synthesis and chemical research, and appears in studies of flavoring agents and tanning processes. The solution form reduces handling of the neat aldehyde; users should follow the supplier safety data sheet for storage, transport, and handling requirements.
  • 40% aqueous solution for simplified handling and reduced vapor hazards.
  • Reactive dicarbonyl reagent for organic synthesis and analytical studies.
  • Applicable to flavoring agent research and tanning process investigations.
  • Molecular weight 72.06 g·mol⁻¹ and formula C3H4O2 aid analytical identification.
  • Refer to the safety data sheet for storage, handling, and hazard information.