Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyruvic aldehyde | 78-98-8 | MFCD00006960 | 97.3% | 72.06 g/mol | C3H4O2 | 5 ML
SDP

Catalog No. 5000299941
Encompass_Preferred

Pyruvic aldehyde (40% in water) is an aqueous solution used as a reagent in organic synthesis, and as an intermediate in flavor and tanning applications. It is intended for research use and should be handled using appropriate laboratory safety procedures; store frozen to preserve stability.

  • Concentration: 40% in water.
  • Molecular formula: C3H4O2; molecular weight: 72.06 g/mol.
  • Reported purity: 97.3%.
  • Supplied as a liquid solution in small laboratory volumes.
  • Storage: keep at -20°C; shelf life up to 2 years.
  • Intended use: for research use only.

Catalog No. 50-002-99941 Supplier Medchemexpress LLC Supplier No. HYW0200145ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyruvic aldehyde (40% in water) is an aqueous solution used as a reagent in organic synthesis, and as an intermediate in flavor and tanning applications. It is intended for research use and should be handled using appropriate laboratory safety procedures; store frozen to preserve stability.

  • Concentration: 40% in water.
  • Molecular formula: C3H4O2; molecular weight: 72.06 g/mol.
  • Reported purity: 97.3%.
  • Supplied as a liquid solution in small laboratory volumes.
  • Storage: keep at -20°C; shelf life up to 2 years.
  • Intended use: for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.