Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyruvonitrile

Catalog No. 26921225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-921-225G 25 g
26-921-25G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-921-225G Supplier Sigma Aldrich Supplier No. 26921225G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • May contain ca
  • 5% mesitylenepyruvonitrile was used in the synthesis of cuprate-carbonyl -complexes by reaction with me2culi in thf

Specifications

CAS 631-57-2
Density 0.974 g/mL (at 25°C (literature))
Boiling Point 92°C to 93°C (lit.)
Molecular Formula C3H3NO
Refractive Index n20/D 1.376 (literature)
Linear Formula CH3COCN
MDL Number MFCD00001838
Quantity 25 g
Synonym 2-Oxopropionitrile; Acetyl cyanide
Molecular Weight (g/mol) 69.06
Percent Purity 90%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.