Learn More
Apexbio Technology LLC Pyrvinium Pamoate, 50mg CAS# 3546-41-6
SDP
Catalog No. 501998434
Encompass_Preferred
Payrvinium pamoate is an ancient anthelmintic used to treat intestinal helminthiasis. As it inhibits the Wnt / -catenin pathway in certain types of cancer, it has attracted attention as an anticancer drug. In vitro, palmitate pyruvate treatment inhibited AKT-mediated phosphorylation of GSK3 at S9 and reduced the protein levels of -catenin and its downstream targets (c-Myc, cyclin D1). In addition, palmitate pyruvate significantly inhibited migration and invasion in vitro. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8434 Supplier Apexbio Technology LLC Supplier No. B839350
Payrvinium pamoate is an ancient anthelmintic used to treat intestinal helminthiasis. As it inhibits the Wnt / -catenin pathway in certain types of cancer, it has attracted attention as an anticancer drug. In vitro, palmitate pyruvate treatment inhibited AKT-mediated phosphorylation of GSK3 at S9 and reduced the protein levels of -catenin and its downstream targets (c-Myc, cyclin D1). In addition, palmitate pyruvate significantly inhibited migration and invasion in vitro. Other sizes are also available. Please inqury us for quote.