Learn More
Medchemexpress LLC HY-U00437 5mg Medchemexpress, Pz-1 CAS:1800505-64-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004375MG
Encompass_Preferred
Medchemexpress, HY-U00437 5mg Pz-1 CAS:1800505-64-9 Pz-1 is a potent RET and VEGFR2 inhibitor with IC 50 s of less than 1 nM for both wild type kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4448