Learn More
Medchemexpress LLC HY-107146 5mg Medchemexpress, PZ-128 CAS:371131-16-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1071465MG
Encompass_Preferred
Medchemexpress, HY-107146 5mg PZ-128 CAS:371131-16-7 PZ-128 (P1pal-7), a cell-penetrating lipopeptide pepducin, is a first-in-class, specific and reversible protease-activated receptor-1 (PAR1) antagonist. PZ-128 targets the cytoplasmic surface of PAR1 and interrupts signaling to internally-located G (PAR1-G) proteins. PZ-128 has antiplatelet, anti-metastatic, anti-angiogenic and anticancer effects[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0698