Learn More
Bachem Pz-Pro-Leu-Gly-Pro-D-Arg-OH
SDP
Catalog No. 50260461
Encompass_Preferred
Pz-Pro-Leu-Gly-Pro-D-Arg-OH Trifluoroacetate, 17011-78-8, C38H52N10O8, Mr 776.89, 1g. Pz-Peptide. Chromogenic substrate for the assay of collagenase and thimet oligopeptidase (Pz-peptidase, metalloendopeptidase 24.15, collagenase-like peptidase). Enzymatic cleavage yields Pz-Pro-Leu-OH (or Pz-Pro-OH). The cleavage can be monitored by analytical HPLC, detection at 320 nm.
Catalog No. 50-260-461 Supplier Bachem Supplier No. 4000833.1000
Add to Cart
Add to Cart
Pz-Pro-Leu-Gly-Pro-D-Arg-OH Trifluoroacetate, 17011-78-8, C38H52N10O8, Mr 776.89, 1g. Pz-Peptide. Chromogenic substrate for the assay of collagenase and thimet oligopeptidase (Pz-peptidase, metalloendopeptidase 24.15, collagenase-like peptidase). Enzymatic cleavage yields Pz-Pro-Leu-OH (or Pz-Pro-OH). The cleavage can be monitored by analytical HPLC, detection at 320 nm.