Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PZ703b hydrochloride | 2471970-56-4 | 99.2% | 1636.90 | 1 MG
SDP

Catalog No. 5000438635
Encompass_Preferred

PZ703b hydrochloride | 2471970-56-4 | 99.2% | 1636.90 | 1 MG

Catalog No. 50-004-38635 Supplier Medchemexpress LLC Supplier No. HY115718B1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PZ703b hydrochloride | 2471970-56-4 | 99.2% | 1636.90 | 1 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.