Learn More
Medchemexpress LLC PZ703b trifluoroacetate | 98.6% | 1714.46 g/mol | C82H103ClF6N10O13S4 | 5 MG
SDP
Catalog No. 5000264363
Encompass_Preferred
PZ703b TFA is a Bcl-xL-targeting PROTAC degrader provided for laboratory research use. It induces rapid BCL-XL degradation, triggers caspase-3-mediated apoptosis, and inhibits cancer cell proliferation in vitro, including activity in bladder cancer models and hematologic cell lines. Supplied as a trifluoroacetate salt, it is intended for biochemical and cell-based studies only.
  • Bcl-xL PROTAC degrader that promotes targeted protein degradation.
  • Induces caspase-3-mediated apoptosis in susceptible cell lines.
  • Inhibits cell proliferation and can act synergistically with other small-molecule inhibitors in vitro.
  • Demonstrated low-nanomolar cellular potency in selected hematologic cell lines.
  • High purity (>98%) suitable for biochemical and cell-based assays.
  • Available in small milligram pack sizes for research use only.
Catalog No. 50-002-64363 Supplier Medchemexpress LLC Supplier No. HY115718A5MG
PZ703b TFA is a Bcl-xL-targeting PROTAC degrader provided for laboratory research use. It induces rapid BCL-XL degradation, triggers caspase-3-mediated apoptosis, and inhibits cancer cell proliferation in vitro, including activity in bladder cancer models and hematologic cell lines. Supplied as a trifluoroacetate salt, it is intended for biochemical and cell-based studies only.
  • Bcl-xL PROTAC degrader that promotes targeted protein degradation.
  • Induces caspase-3-mediated apoptosis in susceptible cell lines.
  • Inhibits cell proliferation and can act synergistically with other small-molecule inhibitors in vitro.
  • Demonstrated low-nanomolar cellular potency in selected hematologic cell lines.
  • High purity (>98%) suitable for biochemical and cell-based assays.
  • Available in small milligram pack sizes for research use only.