Learn More
PROTIDE PHARMACEUTICAL pZerve Cryopreservation Solution, 20 mL
Catalog No. NC0940390
Encompass
A unique cryopreservation solution that does not contain DMSO, fetal bovine serum, or animal protein. pZerve is ready to use and does not require any dilution or further processing.
Catalog No. NC0940390 Supplier PROTIDE PHARMACEUTICAL Supplier No. 5720
A unique cryopreservation solution that does not contain DMSO, fetal bovine serum, or animal protein. pZerve is ready to use and does not require any dilution or further processing.