Learn More
Sigma Aldrich Fine Chemicals Biosciences pZerveTM Cryopreservation
Catalog No. 502905855
Encompass
pZerve is a cryopreservation SOLUTIONon that does not contain dimethyl sulfoxide (DMSO) fetal bovine serum or other animal protein. Optimal recovery is achieved through elimination of these harmful components. pZerve is ready to use does not require any dilution or further processing and can be used for cells cultured in serum free medium or with serum containing medium.
Catalog No. 50-290-5855 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z165360ML
pZerve is a cryopreservation SOLUTIONon that does not contain dimethyl sulfoxide (DMSO) fetal bovine serum or other animal protein. Optimal recovery is achieved through elimination of these harmful components. pZerve is ready to use does not require any dilution or further processing and can be used for cells cultured in serum free medium or with serum containing medium.