Learn More
TARGETMOL CHEMICALS INC PZM21 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T438610MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PZM21 is an effective and selective u opioid receptor agonist (EC50 1.8 nM). purity: 99%
Catalog No. 50-257-9891