Learn More
World Precision Instrument PZMIII Wide Field 20x Eyepieces (pair)
SDP
Catalog No. 502521426
Encompass
  • Microscope eyepieces for the PZMIII microscope
  • One pair of eyepieces
  • 20x magnification
Catalog No. 50-252-1426 Supplier World Precision Instrument Supplier No. 501371
  • Microscope eyepieces for the PZMIII microscope
  • One pair of eyepieces
  • 20x magnification