Learn More
World Precision Instrument PZMIV 0.50x, Planachromatic Objective - 187 mm WD
SDP
Catalog No. 502521343
Encompass
  • Objective for the PZMIV series microscopes
  • Planachromat microscope objective
  • 0.5x magnification
  • 187 mm working distance
Catalog No. 50-252-1343 Supplier World Precision Instrument Supplier No. 502017
  • Objective for the PZMIV series microscopes
  • Planachromat microscope objective
  • 0.5x magnification
  • 187 mm working distance