Learn More
World Precision Instrument PZMIV Binocular Body
SDP
Catalog No. 502521356
Encompass
The fourth generation of WPI’s precision stereo zoom microscopes uses modular, two parallel beam path design and high quality optical system. The advanced optical design with planachromatic objectives provides sharp and distortion-free contrast image throughout the entire zoom range and comes with an impressive 5-year warranty. The microscope offers planachromatic objectives the ensure no optical distortion. It has a large zoom ratio of 8:1, and a large zoom range of 0.62x-5.0x. It has step and continuous zoom and comes with a 5-year warranty.
Catalog No. 50-252-1356 Supplier World Precision Instrument Supplier No. 502000
The fourth generation of WPI’s precision stereo zoom microscopes uses modular, two parallel beam path design and high quality optical system. The advanced optical design with planachromatic objectives provides sharp and distortion-free contrast image throughout the entire zoom range and comes with an impressive 5-year warranty. The microscope offers planachromatic objectives the ensure no optical distortion. It has a large zoom ratio of 8:1, and a large zoom range of 0.62x-5.0x. It has step and continuous zoom and comes with a 5-year warranty.