Learn More
World Precision Instrument PZMTIII Trinocular Body
SDP
Catalog No. 502521419
Encompass
Quality and precision to improve your vision
This microscope offers stereo viewing with ample working distance when used with the long working distance (LWD) option. The long working distance (LWD) option with the 501371 and 501375 on a trinocular microscope shows nearly the same scene on the screen as the viewer sees in the eyepieces.
Catalog No. 50-252-1419 Supplier World Precision Instrument Supplier No. 501379
Quality and precision to improve your vision
This microscope offers stereo viewing with ample working distance when used with the long working distance (LWD) option. The long working distance (LWD) option with the 501371 and 501375 on a trinocular microscope shows nearly the same scene on the screen as the viewer sees in the eyepieces.