Learn More
World Precision Instrument PZMTIV Trinocular Body
SDP
Catalog No. 502521355
Encompass
Modular, two parallel beam path design with high quality optics
The fourth generation of WPI’s trinocular precision stereo zoom microscopes uses modular, two parallel beam path design and high quality optical system. This trinocular microscope is a true trinocular, with continuous operation of both eyepieces and photo tube simultaneously. There is no need to block the right eyepiece to use the photoport.
The advanced optical design with planachromatic objectives provides sharp and distortion-free contrast image throughout the entire zoom range and comes with an impressive 5-year warranty.
Catalog No. 50-252-1355 Supplier World Precision Instrument Supplier No. 502001
Modular, two parallel beam path design with high quality optics
The fourth generation of WPI’s trinocular precision stereo zoom microscopes uses modular, two parallel beam path design and high quality optical system. This trinocular microscope is a true trinocular, with continuous operation of both eyepieces and photo tube simultaneously. There is no need to block the right eyepiece to use the photoport.
The advanced optical design with planachromatic objectives provides sharp and distortion-free contrast image throughout the entire zoom range and comes with an impressive 5-year warranty.