Learn More
MSC Spraying Systems Co. Nozzle, Stainless Steel, Flat Spray, 1 ea
Catalog No. 502817318
Encompass
This nozzle delivers high-impact sprays and high-pressure operation for optimal cleaning, making it ideal for pressure washing. Its optimized design incorporates an internal guide vane to improve fluid dynamics, minimize turbulence, and enhance cleaning performance. Constructed from specially hardened 404 series stainless steel, it offers extended wear life and precise flow control.
  • Provides high-impact sprays and high-pressure operation for effective cleaning
  • Optimized design with internal guide vane minimizes turbulence for superior cleaning
  • Hardened 404 series stainless steel construction ensures longer wear life and flow control accuracy
  • Delivers an even edge flat spray pattern
  • Operates at pressures from 40 to 3000 psi
  • Achieves uniform spray distribution from 0.7 to 6.1 gpm
  • Features a 25° spray angle
  • Equipped with a 1/4 in Male BSPT inlet connection
Catalog No. 50-281-7318 Supplier MSC Supplier No. 02925071
This nozzle delivers high-impact sprays and high-pressure operation for optimal cleaning, making it ideal for pressure washing. Its optimized design incorporates an internal guide vane to improve fluid dynamics, minimize turbulence, and enhance cleaning performance. Constructed from specially hardened 404 series stainless steel, it offers extended wear life and precise flow control.
  • Provides high-impact sprays and high-pressure operation for effective cleaning
  • Optimized design with internal guide vane minimizes turbulence for superior cleaning
  • Hardened 404 series stainless steel construction ensures longer wear life and flow control accuracy
  • Delivers an even edge flat spray pattern
  • Operates at pressures from 40 to 3000 psi
  • Achieves uniform spray distribution from 0.7 to 6.1 gpm
  • Features a 25° spray angle
  • Equipped with a 1/4 in Male BSPT inlet connection