Learn More
Bioworld Q-Cons™ Rapid Protein Concentrators, 25mL Spin Columns, 25 PK(s)
Catalog No. 50253075
Encompass_Preferred
Quick Concentrators™ for proteins. Prepacked columns of Q-Cons™- just add your dilute protein and spin, then collect concentrated sample. Concentrate proteins 5 to 20 fold in 5 minutes.Specifications:•25mL column•Concentrates 25 -30 mlvolume
Catalog No. 50-253-075 Supplier Bioworld Supplier No. 220400632
Quick Concentrators™ for proteins. Prepacked columns of Q-Cons™- just add your dilute protein and spin, then collect concentrated sample. Concentrate proteins 5 to 20 fold in 5 minutes.Specifications:•25mL column•Concentrates 25 -30 mlvolume