Learn More
Bioworld Q-Cons™ Rapid Protein Concentrators, 5mL Spin Columns, 10 PK(s)
Catalog No. 50253072
Encompass_Preferred
Quick Concentrators™ for proteins. Prepacked columns of Q-Cons™- just add your dilute protein and spin, then collect concentrated sample. Concentrate proteins 5 to 20 fold in 5 minutes.Specifications:•5ml column•Concentrates 4 -5 ml volume
Catalog No. 50-253-072 Supplier Bioworld Supplier No. 220400621
Quick Concentrators™ for proteins. Prepacked columns of Q-Cons™- just add your dilute protein and spin, then collect concentrated sample. Concentrate proteins 5 to 20 fold in 5 minutes.Specifications:•5ml column•Concentrates 4 -5 ml volume