Learn More
OraSure Technologies Q.E.D.™ Saliva Alcohol Test, A-150
SDP
Catalog No. 23222108 Shop All OraSure Technologies Products
Change view
Click to view available options
Quantity:
30 Tests
Catalog No. Quantity
23-222-108 30 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-222-108 Supplier OraSure Technologies Supplier No. 31150B
This item is not returnable. View return policy
The Q.E.D.™ Saliva Alcohol Test is the 'on-the-spot leader' in the rapid, accurate and quantitative determination of alcohol in saliva. With a high correlation to blood-alcohol levels, Q.E.D. has a testing range of 0-145 mg/dL and provides quantitative results in minutes.
  • Reliable, Fast, Simple
  • Non-invasive
  • Quantitative results in minutes
  • Reads like a thermometer
  • No instrumentation
  • Long shelf life
  • Built-in QA Spot™ assures quality control for every test
Specifications
Certifications/Compliance CLIA-waived and DOT-approved alcohol testing
Type Saliva Alcohol Test
For Use With (Application) On-Site, Low-Cost Alternative to Breath or Blood Alcohol Testing
Quantity 30 Tests
Test Time <5 min.