Learn More
Sigma Aldrich Fine Chemicals Biosciences Q SepharoseR Fast Flow Cyt
Catalog No. 502907607
Encompass
Q Sepharose(R) Fast Flow is a strong anion exchanger based on the well established Sepharose(R) Fast Flow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.Q Sepharose(R) Fast Flow is part of the Sepharose(R) Fast Flow ion exchange platform which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads with quaternary ammonium (Q) strong anion exchange groups. Q Sepharose(R) Fast Flow has high chemical stability allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with Q Sepharose(R) Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools.As member of the BioProcess media range Q Sepharose(R) Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Catalog No. 50-290-7607 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17051001
Q Sepharose(R) Fast Flow is a strong anion exchanger based on the well established Sepharose(R) Fast Flow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.Q Sepharose(R) Fast Flow is part of the Sepharose(R) Fast Flow ion exchange platform which has been the industrial standard for ion exchange chromatography during recent decades. It is composed of crosslinked 6% agarose beads with quaternary ammonium (Q) strong anion exchange groups. Q Sepharose(R) Fast Flow has high chemical stability allowing well proven cleaning-in-place (CIP) and sanitization protocols. Scale-up with Q Sepharose(R) Fast Flow is straight forward and the medium is available in a range of pre-packed process development tools.As member of the BioProcess media range Q Sepharose(R) Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.