Learn More
Sigma Aldrich Fine Chemicals Biosciences Q SepharoseR fast flow Cyt
Catalog No. 502901608
Encompass
Q Sepharose(R) fast fLow is a strong anion exchanger based on the well established Sepharose(R) fast fLow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.
Catalog No. 50-290-1608 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17051010
Q Sepharose(R) fast fLow is a strong anion exchanger based on the well established Sepharose(R) fast fLow ion exchange platform extensively used for preparative protein separations in both research and industrial applications.