Learn More
Sigma Aldrich Fine Chemicals Biosciences Q SepharoseR High Performa
Catalog No. 502904587
Encompass
Q Sepharose(R) High Performance is a strong anion exchange BioProcess medium with high reSOLUTIONon designed for high intermediate and polishing steps in downstream processing.
Catalog No. 50-290-4587 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17101401
Q Sepharose(R) High Performance is a strong anion exchange BioProcess medium with high reSOLUTIONon designed for high intermediate and polishing steps in downstream processing.