Learn More
Sigma Aldrich Fine Chemicals Biosciences Q SepharoseR Xl Cytiva 17
Catalog No. 502908107
Encompass
Q Sepharose(R) XL is a strong anion exchanger designed for use in the capture steps of biomolecules from clarified feed-stocks.Q Sepharose(R) XL is based on a highly cross-linked bead formed 6% agarose matrix similar to the well established Q Sepharose(R) Fast Flow media. Dextran chains are covalently coupled to the agarose matrix and modified with quaternary ammonium (Q) strong anion exchange groups through chemically stable ether bonds. The stable agarose matrix and long flexible chains of dextran with bound charged groups work together to increase loading capacity whilst allowing high flow rates.With its high loading capacities and excellent physical and chemical stability Q Sepharose(R) XL is suitable for the capture stage of a downstream process which is characterised by large loads of crude samples.As member of the BioProcess media range Q Sepharose(R) XL meets industrial demands with security of supply and comprehensive technical and regulatory support.
Catalog No. 50-290-8107 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17507201
Q Sepharose(R) XL is a strong anion exchanger designed for use in the capture steps of biomolecules from clarified feed-stocks.Q Sepharose(R) XL is based on a highly cross-linked bead formed 6% agarose matrix similar to the well established Q Sepharose(R) Fast Flow media. Dextran chains are covalently coupled to the agarose matrix and modified with quaternary ammonium (Q) strong anion exchange groups through chemically stable ether bonds. The stable agarose matrix and long flexible chains of dextran with bound charged groups work together to increase loading capacity whilst allowing high flow rates.With its high loading capacities and excellent physical and chemical stability Q Sepharose(R) XL is suitable for the capture stage of a downstream process which is characterised by large loads of crude samples.As member of the BioProcess media range Q Sepharose(R) XL meets industrial demands with security of supply and comprehensive technical and regulatory support.